Acid red 114 ≥80.0% (by HPLC)

Leverandør: TCI
Warning

A1874-25G
TCIAA1874-25GEA 435 NOK
TCIAA1874-25G
Acid red 114 ≥80.0% (by HPLC)
Acid red 114
Formel: C₃₇H₃₀N₄O₁₀S₃
MDL nummer: MFCD00003894
CAS-nummer: 6459-94-5
EINECS: 229-272-0
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 80.0 area%
Lambda max. 513.0 to 517.0 nm (EtOH (50%))
Absorbance (E1%1cm) min. 300 (EtOH (50%), 513.0 to 517.0nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR