9-Aminocamptothecin 95,0 HPLC_ASSAY_METHOD

Leverandør: TCI

A2063-10MG
TCIAA2063-10MGEA 4025 NOK
TCIAA2063-10MG
9-Aminocamptothecin 95,0 HPLC_ASSAY_METHOD
9-Aminocamptothecin
Formel: C₂₀H₁₇N₃O₄
MW: 363.37 g/mol
MDL nummer: MFCD00909855
CAS-nummer: 91421-43-1
Merck Indeks: 14,00431

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR