Azathioprine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

A2069-25G A2069-5G
TCIAA2069-25GEA 8800 NOK
TCIAA2069-25G TCIAA2069-5G
Azathioprine ≥98.0% (by HPLC, titration analysis)
Azathioprine
Formel: C₉H₇N₇O₂S
Smeltepunkt: 244°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00069203
CAS-nummer: 446-86-6
EINECS: 207-175-4
Merck Indeks: 14,00902

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR