4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)

Leverandør: TCI

A2121-5G
TCIAA2121-5GEA 1225 NOK
TCIAA2121-5G
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₈H₁₅NO₂
Smeltepunkt: 233°C
MDL nummer: MFCD00064951
CAS-nummer: 701-54-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 95.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR