cis-3-Aminocyclohexanecarboxylic acid ≥96.0% (by GC, titration analysis)

Leverandør: TCI

A2127-25G A2127-5G
TCIAA2127-25GEA 6175 NOK
TCIAA2127-25G TCIAA2127-5G
cis-3-Aminocyclohexanecarboxylic acid ≥96.0% (by GC, titration analysis)
cis-3-Aminocyclohexanecarboxylic acid
Formel: C₇H₁₃NO₂
Smeltepunkt: 278°C
MDL nummer: MFCD09753647
CAS-nummer: 16636-51-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR