Allyl-2,2,2-Trichloroacetimidate ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Allyl 2,2,2-trichloroacetimidate, 2,2,2-Trichloroacetimidic Acid Allyl Ester

A2186-5G
TCIAA2186-5GEA 1875 NOK
TCIAA2186-5G
Allyl-2,2,2-Trichloroacetimidate ≥98.0% (by GC)
Allyl-2,2,2-Trichloroacetimidate
Formel: C₅H₆Cl₃NO
MW: 202,46 g/mol
Kokepunkt: 74°C
Flash punkt: 74°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00190710
CAS-nummer: 51479-73-3

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR