[2-(Aminomethyl)phenyl]acetic acid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: [2-(Aminomethyl)phenyl]acetic Acid, 2-Aminomethylphenylacetic acid, 2-(2-(Aminomethyl)phenyl)acetic acid

A2199-5G
TCIAA2199-5GEA 1875 NOK
TCIAA2199-5G
[2-(Aminomethyl)phenyl]acetic acid ≥97.0% (by HPLC, titration analysis)
[2-(Aminomethyl)phenyl]acetic acid
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
MDL nummer: MFCD04038440
CAS-nummer: 40851-65-8
EINECS: 255-110-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR