5-Aza-2'-deoxycytidine

Leverandør: TCI
Warning

Synonymer: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

A2232-20MG A2232-100MG
TCIAA2232-20MGEA 745 NOK
TCIAA2232-20MG TCIAA2232-100MG
5-Aza-2'-deoxycytidine
5-Aza-2'-deoxycytidine
Formel: C₈H₁₂N₄O₄
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00043011
CAS-nummer: 2353-33-5
EINECS: 219-089-4
Merck Indeks: 14,02853

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Elemental analysis (Nitrogen) 23.8 to 25.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR