Anastrozole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: alpha,alpha,alpha',alpha'-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile

A2370-100MG
TCIAA2370-100MGEA 260 NOK
TCIAA2370-100MG
Anastrozole ≥98.0% (by HPLC, titration analysis)
Anastrozole
Formel: C₁₇H₁₉N₅
MDL nummer: MFCD00866298
CAS-nummer: 120511-73-1
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR