5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine

A2528-50MG
TCIAA2528-50MGEA 635 NOK
TCIAA2528-50MG
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98.0% (by HPLC)
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Formel: C₉H₁₄N₄O₅
MW: 258.23 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00869751
CAS-nummer: 2627-69-2
EINECS: 220-097-5
Merck Indeks: 14,00016

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 21.20 to 22.00 %
Specific rotation [a]20/D -61.0 to -66.0 deg (C=1、H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR