AOL-biotin conjugate

Leverandør: TCI

A2659-1ML
TCIAA2659-1MLEA 4550 NOK
TCIAA2659-1ML
AOL-biotin conjugate
AOL-biotin conjugate

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Molecular weight 30 to 40 kDa
Suitability for use in ELISA tests min. 1.0 (human Lactoferrin, 10 micro g/mL, OD450)
Fucose inhibition test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR