Biotin-C5-Amine ≥95.0% (by HPLC)

Leverandør: TCI

A3155-100MG A3155-25MG
TCIAA3155-100MGEA 4575 NOK
TCIAA3155-100MG TCIAA3155-25MG
Biotin-C5-Amine ≥95.0% (by HPLC)
Biotin-C5-Amine
Formel: C₁₅H₂₈N₄O₂S
Smeltepunkt: 122°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00216002
CAS-nummer: 115416-38-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 95.0 area%
Melting point 122.0 to 126.0 deg-C
Specific rotation [a]20/D +40.0 to +44.0 deg (C=0.03, MeOH)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR