Farnesyl thiosalicylic acid ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

A3351-100MG A3351-25MG
TCIAA3351-100MGEA 2550 NOK
TCIAA3351-100MG TCIAA3351-25MG
Farnesyl thiosalicylic acid ≥96.0% (by HPLC, titration analysis)
Farnesyl thiosalicylic acid
Formel: C₂₂H₃₀O₂S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00467723
CAS-nummer: 162520-00-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR