Oxo[5,10,15,20-tetra(4-pyridyl)porphyrinato]titanium(IV) ≥90.0% (by HPLC) determination of glucose in serum and urine

Leverandør: TCI

A5020-100MG
TCIAA5020-100MGEA 7100 NOK
TCIAA5020-100MG
Oxo[5,10,15,20-tetra(4-pyridyl)porphyrinato]titanium(IV) ≥90.0% (by HPLC) determination of glucose in serum and urine
Oxo[5,10,15,20-tetra(4-pyridyl)porphyrinato]titanium(IV)
Formel: C₄₀H₂₄N₈OTi
MW: 680.55 g/mol
MDL nummer: MFCD00191476
CAS-nummer: 105250-49-5

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 90.0 area%
Functionality test to pass test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR