Rhodamin B ≥98.0% (by titrimetric analysis) ion association reagent for photometric and fluorimetric analysis

Leverandør: TCI
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

A5102-1G
TCIAA5102-1GEA 405 NOK
TCIAA5102-1G
Rhodamin B ≥98.0% (by titrimetric analysis) ion association reagent for photometric and fluorimetric analysis
Rhodamin B
Formel: C₂₈H₃₁ClN₂O₃
Smeltepunkt: 199…201 °C
Tetthet: 1,31 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
EINECS: 201-383-9
Merck Indeks: 14,08183
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10372e03d516ef079aaa6a0d6e2b5414d6cf8b20&vid=404adca93b6b79f3cddd12065f2e89f0e455f5b5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Lambda max. 552.0 to 556.0 nm (H2O)
Absorbance (E1%1cm) min. 2,000 (H2O, 552.0 to 556.0 nm)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR