Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

A5160-5G
TCIAA5160-5GEA 350 NOK
TCIAA5160-5G
Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)
Benzyldimethyltetradecylammonium chloride hydrate

Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals.

Formel: C₂₃H₄₂ClN·H₂O
MDL nummer: MFCD00011771
CAS-nummer: 139-08-2
EINECS: 205-352-0
FN: 2923
ADR: 8,III
Merck Indeks: 14,01059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=142cea45fee7a754ef2b840da7118d4e251d287e&vid=145d7ff51e90694622f785b1dee98fc83e8a4941&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (TPBNa Method) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 12.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR