MDEPAP [=1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide] ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide

A5400-1G
TCIAA5400-1GEA 1425 NOK
TCIAA5400-1G
MDEPAP [=1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide] ≥95.0% (by HPLC, total nitrogen)
MDEPAP [=1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide]

Extraction-spectrophotometric reagent for anionic surfactants

Formel: C₁₆H₂₁IN₄
MW: 396,28 g/mol
MDL nummer: MFCD00082424
CAS-nummer: 74920-80-2

Bestill nå

Spesifikasjon Testresultater

Appearance Light gray to Brown to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 186.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR