N-Succinimidyl 4-Nitrophenylacetate ≥98.0% (by total nitrogen basis) for HPLC labeling

Leverandør: TCI
Warning

Synonymer: N-(4-Nitrophenylacetoxy)succinimide

A5522-1G
TCIAA5522-1GEA 825 NOK
TCIAA5522-1G
N-Succinimidyl 4-Nitrophenylacetate ≥98.0% (by total nitrogen basis) for HPLC labeling
N-Succinimidyl 4-Nitrophenylacetate
Formel: C₁₂H₁₀N₂O₆
MW: 278.22 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00047124
CAS-nummer: 68123-33-1
EINECS: 268-551-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 181.0 to 185.0 deg-C
Solubility in hot Acetone almost transparency
Effect test of HPLC derivatization Effective as labeling agent for Diethylamine.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR