O-(4-Nitrobenzyl)hydroxylamine hydrochloride 99,0 HPLC_ASSAY_METHOD HPLC Labeling

Leverandør: TCI

Synonymer: O-(4-Nitrobenzyl)-hydroxylammonium chloride

A5532-1G A5532-5G
TCIAA5532-1GEA 770 NOK
TCIAA5532-1G TCIAA5532-5G
O-(4-Nitrobenzyl)hydroxylamine hydrochloride 99,0 HPLC_ASSAY_METHOD HPLC Labeling
O-(4-Nitrobenzyl)hydroxylamine hydrochloride
Formel: C₇H₉ClN₂O₃
MW: 204.61 g/mol
MDL nummer: MFCD00012954
CAS-nummer: 2086-26-2
EINECS: 218-228-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5b736caa18d16c485db0e49f5f40094a15dab9&vid=670d3dab143bf732c4286826f7ed8c35d3680e74&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot Water almost transparency
Effect test of HPLC derivatization Effective as labeling agent for Valeraldehyde


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR