syn-Benzaldehyde oxime ≥90.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (E)-Benzaldehyde oxime, alpha-Benzaldoxime

B0011-25G B0011-500G B0011-100G
TCIAB0011-25GEA 660 NOK
TCIAB0011-25G TCIAB0011-500G TCIAB0011-100G
syn-Benzaldehyde oxime ≥90.0% (by GC)
syn-Benzaldehyde oxime
Formel: C₇H₇NO
Kokepunkt: 123°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00002119
CAS-nummer: 622-31-1
EINECS: 213-261-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb290ae1f4c2f8aaa445b6b2c61896f06d5163b1&vid=bb290ae1f4c2f8aaa445b6b2c61896f06d5163b1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR