3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: m-Tolyltetrazolium Red Formazan

B0283-100MG
TCIAB0283-100MGEA 715 NOK
TCIAB0283-100MG
3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (by HPLC)
3,5-Diphenyl-1-(m-tolyl)formazan
Formel: C₂₀H₁₈N₄
MDL nummer: MFCD00059314
CAS-nummer: 71274-65-2

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 154.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR