2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium chloride ≥80.0% (by titrimetric analysis)

Leverandør: TCI

B0339-100MG
TCIAB0339-100MGEA 915 NOK
TCIAB0339-100MG
2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium chloride ≥80.0% (by titrimetric analysis)
2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium chloride
Formel: C₂₀H₁₇ClN₄O
MDL nummer: MFCD00060032
CAS-nummer: 10560-45-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 80.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR