Dibenzyl disulphide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Benzyl disulphide

B0417-25G
TCIAB0417-25GEA 525 NOK
TCIAB0417-25G
Dibenzyl disulphide ≥98.0% (by GC)
Dibenzyl disulphide
Formel: C₁₄H₁₄S₂
MW: 246,4 g/mol
Kokepunkt: 142 - 148 °C (0,05 -rr)
Smeltepunkt: 62…67 °C
Tetthet: 1,302 g/cm³ (25 °C)
MDL nummer: MFCD00004783
CAS-nummer: 150-60-7
EINECS: 205-764-0
Merck Indeks: 14,03013
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fcca2fca2c8b85bcc1bea493445f12d5dde494a&vid=3fcca2fca2c8b85bcc1bea493445f12d5dde494a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 73.0 deg-C
Solubility in hot Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR