N,O-Bis(trimetylsilyl)acetamid 25% in acetonitrile trimethylsilylating reagent, for NH₂ compounds

Leverandør: TCI
Danger

B0510-12ML
TCIAB0510-12MLEA 310 NOK
TCIAB0510-12ML
N,O-Bis(trimetylsilyl)acetamid 25% in acetonitrile trimethylsilylating reagent, for NH₂ compounds
N,O-Bis(trimetylsilyl)acetamid
Formel: C₈H₂₁NOSi₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008270
CAS-nummer: 10416-59-8
EINECS: 233-892-7
FN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1005f7d84336f89772ac9abdb5cf250c3fe65ad&vid=4c01c12c82bc4baef251bed014de3f68f39fbd8d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Effect test of TMS derivatization Effective as derivatization agent DL-Threonine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR