L(-)-Bornyl acetate ≥70% (by GC) (contains ca. 20% Isobornyl Acetate)

Leverandør: TCI

B0526-25ML
TCIAB0526-25MLEA 220 NOK
TCIAB0526-25ML
L(-)-Bornyl acetate ≥70% (by GC) (contains ca. 20% Isobornyl Acetate)
L(-)-Bornyl acetate
Formel: C₁₂H₂₀O₂
Flash punkt: 84°C
MDL nummer: MFCD00135944
CAS-nummer: 5655-61-8
EINECS: 227-101-4
Merck Indeks: 14,01339

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 70.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR