Bordeaux red

Leverandør: TCI
Warning

B0779-25G
TCIAB0779-25GEA 700 NOK
TCIAB0779-25G
Bordeaux red
Bordeaux red
Formel: C₂₀H₁₄N₂O₇S₂
MDL nummer: MFCD00003887
CAS-nummer: 5858-33-3
EINECS: 227-489-5

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Lambda max. 521.0 to 525.0 nm (H2O)
Absorbance (E1%1cm) min. 250 (H2O, 521.0 to 525.0 nm)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR