2,2'-Bi-4-lepidine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 4,4'-Dimethyl-2,2'-biquinoline, 2,2'-Di-4-lepidyl

B0841-100MG
TCIAB0841-100MGEA 2875 NOK
TCIAB0841-100MG
2,2'-Bi-4-lepidine ≥98.0% (by titrimetric analysis)
2,2'-Bi-4-lepidine
Formel: C₂₀H₁₆N₂
MW: 284.36 g/mol
MDL nummer: MFCD00059753
CAS-nummer: 7654-51-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR