4-Butoksybenso syre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

B0896-25G
TCIAB0896-25GEA 690 NOK
TCIAB0896-25G
4-Butoksybenso syre ≥98.0% (by GC, titration analysis)
4-Butoksybenso syre
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Kokepunkt: 319
Smeltepunkt: 149
MDL nummer: MFCD00002546
CAS-nummer: 1498-96-0
EINECS: 216-104-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ab1be14a064faf42053c18bb484b76c5afbed2&vid=32ab1be14a064faf42053c18bb484b76c5afbed2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR