1,2-Bis(difenylfosfino)etan ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

B1137-10G B1137-25G
TCIAB1137-10GEA 730 NOK
TCIAB1137-10G TCIAB1137-25G
1,2-Bis(difenylfosfino)etan ≥97.0% (by GC, titration analysis)
1,2-Bis(difenylfosfino)etan
Formel: C₂₆H₂₄P₂
MW: 398.42 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003047
CAS-nummer: 1663-45-2
EINECS: 216-769-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859b79caed6a986beb95f628fa03feeffd7ed0a1&vid=859b79caed6a986beb95f628fa03feeffd7ed0a1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 137.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR