1,3-Bis(difenylfosfino)propan ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

B1138-25G B1138-5G
TCIAB1138-25GEA 1100 NOK
TCIAB1138-25G TCIAB1138-5G
1,3-Bis(difenylfosfino)propan ≥98.0% (by GC)
1,3-Bis(difenylfosfino)propan
Formel: C₂₇H₂₆P₂
MW: 412.45 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003050
CAS-nummer: 6737-42-4
EINECS: 229-791-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2fd0ab48e0704cc2742e0bd31418a5775b2ba70&vid=b2fd0ab48e0704cc2742e0bd31418a5775b2ba70&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystaline
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 96.0 %
Melting point 58.0 to 63.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR