2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole ≥97.0% (by titrimetric analysis) photopolymerization initiator

Leverandør: TCI

Synonymer: 2,2-Bis(2-chlorophenyl)-4,4,5,5-tetraphenyl-1,2-biimidazole

B1225-25G
TCIAB1225-25GEA 930 NOK
TCIAB1225-25G
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole ≥97.0% (by titrimetric analysis) photopolymerization initiator
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole
Formel: C₄₂H₂₈Cl₂N₄
MW: 659.61 g/mol
MDL nummer: MFCD00060081
CAS-nummer: 7189-82-4
EINECS: 230-555-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 196.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR