4-Bromo-4'-heptylbiphenyl ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 4-Bromo-4'-heptyl-1,1'-biphenyl

B1304-5G
TCIAB1304-5GEA 1750 NOK
TCIAB1304-5G
4-Bromo-4'-heptylbiphenyl ≥97.0% (by GC)
4-Bromo-4'-heptylbiphenyl
Formel: C₁₉H₂₃Br
MW: 331,2899 g/mol
MDL nummer: MFCD00060107
CAS-nummer: 58573-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ee0ad32fa912f0975df9ea966411ca0661bc488&vid=3ee0ad32fa912f0975df9ea966411ca0661bc488&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 92.0 to 95.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR