Bis[2-(4-methoxyphenoxy)ethyl]ether ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4,4'-((Oxybis(ethane-2,1-diyl))bis(oxy))bis(methoxybenzene)

B1479-1G
TCIAB1479-1GEA 1075 NOK
TCIAB1479-1G
Bis[2-(4-methoxyphenoxy)ethyl]ether ≥98.0% (by GC)
Bis[2-(4-methoxyphenoxy)ethyl]ether
Formel: C₁₈H₂₂O₅
MW: 318,37 g/mol
MDL nummer: MFCD00143755
CAS-nummer: 104104-12-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 102.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR