Benzyl-2,2,2-trichloroacetimidate ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Trichloroacetimidic Acid Benzyl Ester, Benzyl 2,2,2-trichloroacetimidate

B1483-25G
TCIAB1483-25GEA 1275 NOK
TCIAB1483-25G
Benzyl-2,2,2-trichloroacetimidate ≥97.0% (by GC)
Benzyl-2,2,2-trichloroacetimidate
Formel: C₉H₈Cl₃NO
MW: 252,52 g/mol
Kokepunkt: 150°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000805
CAS-nummer: 81927-55-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR