Ethyl-2-bromobenzoate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Ethyl 2-bromobenzoate, 2-Bromobenzoic acid ethyl ester

B1514-25G
TCIAB1514-25GEA 650 NOK
TCIAB1514-25G
Ethyl-2-bromobenzoate ≥98.0% (by GC)
Ethyl-2-bromobenzoate
Formel: C₉H₉BrO₂
MW: 229,07 g/mol
Kokepunkt: 255°C
Tetthet: 1,441 g/cm³ (20 °C)
MDL nummer: MFCD00015443
CAS-nummer: 6091-64-1
EINECS: 228-034-3

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR