Belleau's reagent sulphurating reagent

Leverandør: TCI

Synonymer: 2,4-Bis(4-phenoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulphide

B1517-5G
TCIAB1517-5GEA 1625 NOK
TCIAB1517-5G
Belleau's reagent sulphurating reagent
Belleau's reagent
Formel: C₂₄H₁₈O₂P₂S₄
MW: 528.59 g/mol
MDL nummer: MFCD00191383
CAS-nummer: 88816-02-8
Merck Indeks: 14,01026
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77a65995846956e8769455243a83ebf99a92b72d&vid=77a65995846956e8769455243a83ebf99a92b72d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Melting point 190.0 to 197.0 deg-C
Performance test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR