Bis(4-hydroxy-3-methylphenyl)sulphide ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4,4'-Thiodi(o-cresol), 4,4'-Thiobis(2-methylphenol)

B1527-25G
TCIAB1527-25GEA 1125 NOK
TCIAB1527-25G
Bis(4-hydroxy-3-methylphenyl)sulphide ≥98.0% (by GC)
Bis(4-hydroxy-3-methylphenyl)sulphide
Formel: C₁₄H₁₄O₂S
MW: 246,32 g/mol
MDL nummer: MFCD00142117
CAS-nummer: 24197-34-0
EINECS: 403-330-7
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 125.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR