Boc-trans-4-hydroxy-L-proline ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline

B1635-5G
TCIAB1635-5GEA 525 NOK
TCIAB1635-5G
Boc-trans-4-hydroxy-L-proline ≥98.0% (by titrimetric analysis)
Boc-trans-4-hydroxy-L-proline
Formel: C₁₀H₁₇NO₅
MDL nummer: MFCD00053370
CAS-nummer: 13726-69-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -67.0 to -69.0 deg (C=1, MeOH)
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR