N-Benzylcinchoninium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst

Leverandør: TCI
Warning

Synonymer: BCNC, N-Benzylcinchonium chloride, (1S,2R,4S,5R)-1-Benzyl-2-(hydroxy(quinolin-4-yl)methyl)-5-vinylquinuclidin-1-ium chloride

B1689-5G
TCIAB1689-5GEA 1225 NOK
TCIAB1689-5G
N-Benzylcinchoninium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
N-Benzylcinchoninium chloride
Formel: C₂₆H₂₉ClN₂O
MW: 420,98 g/mol
MDL nummer: MFCD00082423
CAS-nummer: 69221-14-3
EINECS: 273-915-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +167 to +174 deg (C=0.4, MeOH:H2O=1:5)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR