2-Ethylhexyl benzoate ≥99.0% (by GC)

Leverandør: TCI

Synonymer: Benzoic acid 2-Ethylhexyl ester

B1977-25ML
TCIAB1977-25MLEA 1350 NOK
TCIAB1977-25ML
2-Ethylhexyl benzoate ≥99.0% (by GC)
2-Ethylhexyl benzoate
Formel: C₁₅H₂₂O₂
MW: 234,34 g/mol
Kokepunkt: 170°C
MDL nummer: MFCD00072620
CAS-nummer: 5444-75-7
EINECS: 226-641-8

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR