4-Bromo-m-toluidine ≥98.0% (by GC)

Leverandør: TCI
Warning

B1990-25G B1990-5G
TCIAB1990-25GEA 660 NOK
TCIAB1990-25G TCIAB1990-5G
4-Bromo-m-toluidine ≥98.0% (by GC)
4-Bromo-m-toluidine
Formel: C₇H₈BrN
Kokepunkt: 240°C
MDL nummer: MFCD00007828
CAS-nummer: 6933-10-4
EINECS: 230-056-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd5d3bfc9057bcdb730836511d0face05853ce3&vid=dfd5d3bfc9057bcdb730836511d0face05853ce3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR