5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde ≥94.0% (by GC)

Leverandør: TCI

Synonymer: 5-Bromo-5''-formyl-2,2':5',2''-terthiophene, 5''-Bromo-[2,2':5',2''-terthiophene]-5-carbaldehyde

B2058-1G B2058-5G
TCIAB2058-1GEA 2800 NOK
TCIAB2058-1G TCIAB2058-5G
5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde ≥94.0% (by GC)
5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde
Formel: C₁₃H₇BrOS₃
MDL nummer: MFCD02093442
CAS-nummer: 161726-69-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 94.0 %
Melting point 158.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR