1,4-Diacetoxy-2-butyne ≥95.0% (by GC)

Leverandør: TCI

Synonymer: But-2-yne-1,4-diyl diacetate, 2-Butyne-1,4-diol Diacetate, Bis(acetoxymethyl)acetylene

B2087-25G
TCIAB2087-25GEA 1250 NOK
TCIAB2087-25G
1,4-Diacetoxy-2-butyne ≥95.0% (by GC)
1,4-Diacetoxy-2-butyne
Formel: C₈H₁₀O₄
Kokepunkt: 235°C
MDL nummer: MFCD00014982
CAS-nummer: 1573-17-7
EINECS: 216-394-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Freezing point 23.0 to 26.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR