(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)

Leverandør: TCI
Warning

Synonymer: (S)-AMAC

B2315-100MG
TCIAB2315-100MGEA 3400 NOK
TCIAB2315-100MG
(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
Formel: C₃₂H₃₈CoN₂O₄
MW: 573.60 g/mol
MDL nummer: MFCD06797062
CAS-nummer: 259259-80-8

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Cobalt min. 10.0 %
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR