N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0% (by HPLC)

Leverandør: TCI
Warning

B3087-1G B3087-5G
TCIAB3087-1GEA 865 NOK
TCIAB3087-1G TCIAB3087-5G
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0% (by HPLC)
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine
Formel: C₃₇H₃₅N₃O₇
Smeltepunkt: 119°C
MDL nummer: MFCD00010114
CAS-nummer: 67219-55-0
EINECS: 266-605-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +56.0 to +60.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR