N4-Benzoyl-2'-deoxycytidine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

B3102-1G B3102-100MG
TCIAB3102-1GEA 1350 NOK
TCIAB3102-1G TCIAB3102-100MG
N4-Benzoyl-2'-deoxycytidine ≥98.0% (by HPLC, total nitrogen)
N4-Benzoyl-2'-deoxycytidine
Formel: C₁₆H₁₇N₃O₅
Smeltepunkt: 207°C
MDL nummer: MFCD00010115
CAS-nummer: 4836-13-9
EINECS: 225-421-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +87.0 to +93.0 deg (C=0.25, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR