tert-Butyl hydroperoxide 70%

Leverandør: TCI
Danger

B3153-100G
TCIAB3153-100GEA 300 NOK
TCIAB3153-100G
tert-Butyl hydroperoxide 70%
tert-Butyl hydroperoxide
Formel: C₄H₁₀O₂
Kokepunkt: 89°C
Smeltepunkt: -3°C
Flash punkt: 43°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00002130
CAS-nummer: 75-91-2
EINECS: 200-915-7
FN: 3109
ADR: 5.2,
Merck Indeks: 14,01570

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Iodometric titration) 65.0 to 71.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR