5-Bromo-5-nitro-1,3-dioxane ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Bromonitrodioxane, Bronidox L

B3156-25G
TCIAB3156-25GEA 1225 NOK
TCIAB3156-25G
5-Bromo-5-nitro-1,3-dioxane ≥98.0% (by GC)
5-Bromo-5-nitro-1,3-dioxane
Formel: C₄H₆BrNO₄
MW: 212,00 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00101855
CAS-nummer: 30007-47-7
EINECS: 250-001-7
FN: 3261
ADR: 8,I

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR