Bis(tri-tert-butylphosphine)platinum(0)

Leverandør: TCI

Synonymer: Bis(tri-t-butylphosphine)platinum (0)

B3162-250MG
TCIAB3162-250MGEA 3025 NOK
TCIAB3162-250MG
Bis(tri-tert-butylphosphine)platinum(0)
Bis(tri-tert-butylphosphine)platinum(0)
Formel: C₂₄H₅₄P₂Pt
MW: 599,73 g/mol
MDL nummer: MFCD04974248
CAS-nummer: 60648-70-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR