Butyltrimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

B3233-5G
TCIAB3233-5GEA 850 NOK
TCIAB3233-5G
Butyltrimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Butyltrimethylammonium bis(trifluoromethanesulphonyl)imide
Formel: C₉H₁₈F₆N₂O₄S₂
MW: 396,36 g/mol
Smeltepunkt: 17°C
MDL nummer: MFCD12761449
CAS-nummer: 258273-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f22bba43f49f35b2793148b580bc21295bd6ea0&vid=7f22bba43f49f35b2793148b580bc21295bd6ea0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR